missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSNK1E (CK1 ε) Recombinant Human Protein
CSNK1E (CK1 epsilon) is a serine/threonine kinase.
1826.00 EUR - 7890.00 EUR
Specifications
Accession Number | NP_001885 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.04% Triton¤ X-100, 4mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 52.3kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15458433
|
Invitrogen™
PR5147A |
100μg |
1826.00 EUR
100µg |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10795433
|
Invitrogen™
PV3988 |
1mg |
7890.00 EUR
1mg |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CSNK1E (CK1 epsilon) is implicated in a wide range of cellular functions, including vesicular trafficking, DNA damage repair, cell cycle progression, and cytokinesis.
Specifications
NP_001885 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.04% Triton¤ X-100, 4mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CK1 Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
52.3kDa | |
RUO - research use only | |
CSNK1E (CK1 ε) | |
Human | |
His Tag | |
Frozen |