missing translation for 'onlineSavingsMsg'
Learn More
Learn More
This item is not returnable.
View return policy
Specifications
Permanent | |
Cryostick | |
Cryogenic use | |
80°C | |
12 mm | |
11 mm |
White | |
Cryogenic Label | |
Thermal printer | |
12 mm | |
11 mm |