Learn More
Fisherbrand™ Cryogenic Dots in Laser Sheets
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Come in an easy to use dispenser box with 20 sheets per pack.
Brand: Fisherbrand™
Additional Details : Weight : 0.31750kg
Description
The labels can be printed on and will not bleed. Perfect for distinguishing amongst various specimens.Specifications
Yellow | |
Non-sterile | |
0.37 in. |
Dot label laser sheet | |
0.5 to 1.5mL tubes | |
9.55 mm |