missing translation for 'onlineSavingsMsg'
Learn More

Crocetin, free acid, >90%, MP Biomedicals™

Catalog Number p-4405614
Click to view available options
Quantity:
1 mg
5 mg
25 mg
This item is not returnable. View return policy
27876-94-4
C20H24O4
328.408
PANKHBYNKQNAHN-MQQNZMFNSA-N
crocetin, trans-crocetin, 8,8'-diapocarotenedioic acid, natural yellow 6, 8,8'-diapocarotene-8,8'-dioic acid, 8,8'-diapo-psi,psi-carotenedioic acid, ccris 7484, unii-20tc155l9c, 2e,4e,6e,8e,10e,12e,14e-2,6,11,15-tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedioic acid, .alpha.-crocetin
5281232
CHEBI:3918
(2E,4E,6E,8E,10E,12E,14E)-2,6,11,15-tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedioic acid
CC(=CC=CC=C(C)C=CC=C(C)C(=O)O)C=CC=C(C)C(=O)O
This item is not returnable. View return policy
27876-94-4
C20H24O4
328.408
PANKHBYNKQNAHN-MQQNZMFNSA-N
crocetin, trans-crocetin, 8,8'-diapocarotenedioic acid, natural yellow 6, 8,8'-diapocarotene-8,8'-dioic acid, 8,8'-diapo-psi,psi-carotenedioic acid, ccris 7484, unii-20tc155l9c, 2e,4e,6e,8e,10e,12e,14e-2,6,11,15-tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedioic acid, .alpha.-crocetin
5281232
CHEBI:3918
(2E,4E,6E,8E,10E,12E,14E)-2,6,11,15-tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedioic acid
CC(=CC=CC=C(C)C=CC=C(C)C(=O)O)C=CC=C(C)C(=O)O
Color Orange
Assay Percent Range >90%
Quantity 1 mg
Formula Weight 328.4
Physical Form Powder
Chemical Name or Material Crocetin, free acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'