Learn More
Cole-Parmer™ CPC™ Quick Disconnect Polypropylene Pipe Adapter Bodies
Quick Disconnect couplings feature a thumb latch for easy assembly
Brand: Cole-Parmer™ 31303-07
Additional Details : Weight : 0.01000kg
Description
- High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately)
- Plug insert into coupling body to connect
- Fitting halves lock into place immediately
- To disconnect fitting halves, simply press the button on the coupling body
- Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged
- All fitting bodies have 316 SS latch springs and EPDM seals
Spécification
Adapter | |
71°C | |
3/4 in. Quick-Disconnect | |
Ethylene Oxide (ETO) or Gamma | |
60 psi |
160°F | |
3/4 in. | |
Quick-Disconnect x NPT(M) Connection | |
Polypropylene |