missing translation for 'onlineSavingsMsg'
Learn More

CPC™ Plastic Quick-Disconnect Fittings

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables tests.

Brand:  CPC™ MPC3301239

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11779749

  • 13.80 EUR / Each


Estimated Shipment
21-04-2021
Add to basket

Description

Description

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables

CPC Series couplings assure secure connections, and are easy to operate—even with gloved hands. They are reusable, yet economical enough to allow disposability.

  • Material: Medical-grade ABS, polycarbonate and polysulfone
  • Tubing Range: 1/4 and 3/8 in. I.D. (6.4 to 9.5mm)
  • Fow Rate: Cv∽5.5 (79.25) max.

  • Ergonomic thumb latch
  • Audible 'click' when connected
  • USP Class VI materials
  • Can be sterilized by autoclave, EtO, e-beam, or gamma
  • Coupling halves can be rotated when connected
  • Tapered barb
  • BSE/TSE free materials

  • Cell culture media packaging and transfer lines
  • Harvest and collection applications
  • Buffer solutions
  • Tube sets
  • Urology drainage/collection systems
  • Hypo/hyperthermia blankets

Specifications

Specifications

USP Class VI, ADCF
300°F
60 psi
Polysulfone w/Platinum Cured Silicone Seal
38 mm
19.05mm
-40°F
25 mm
Amber
149°C
Autoclavable
1.4 in.
Sanitary Non-valved Coupling Body
Cell culture media packaging and transfer lines, harvest and collection applications
-40°C
Videos
SDS
Documents