Learn More
Thermo Scientific™ Filtration Hardware, Accessories, and Spare Parts
Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in a wide range of applications and in conjunction with our various lines of filtration devices.
Brand: Thermo Scientific™ 710315-0003
Additional Details : Weight : 0.04500kg
Description
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
These are replacement parts and accessories for Nalgene™ Reusable Filter Funnel with Clamp, Cat #DS0315-0047
The Reusable Filter Funnel Features:
- Tight fit prevents bypass even after autoclaving
- Reservoir graduated from 100 to 250mL in 25mL increments
- Aluminum clamp allows one-handed operation and easy membrane replacement
Includes: Rugged polysulfone funnel; polypropylene copolymer cover with three ports, three friction-fit TPE caps; support plate for 47mm membrane; silicone gasket; No. 8 stopper (0.5625 in. hole); aluminum clamp
Specifications
Cover with 3 TPE Caps | |
Nalgene Reusable Filter Funnel with Clamp | |
Accessory |