Learn More
Corning™ HYPERStack™ Accessories: Stack Manipulator
Accessories for use with Corning HYPERStack products
Brand: Corning™ 11000
Additional Details : Weight : 58.96696kg
Description
- Made of stainless steel for durability
- Accommodates both HYPERStack-36 and CellSTACK-10 vessels
- Ergonomic design
- Easy dials for protocol development
- Handle two units simultaneously or one unit at a time
Specifications
Stainless Steel | |
1/Cs. | |
Stack manipulator |
Hyperstack™ 110and Hyperstack™ 36 Vessels | |
1/Pk. |