Learn More
Corning™ Fiberglass Prefilters
For use in Corning bottle top and vacuum filtration systems.
Brand:  Corning™ 431410
Additional Details : Weight : 0.04536kg
 View more versions of this product
Product Code. 11343775
  • 81.17 EUR / Pack of 100
Estimated Shipment: 29-07-2024
Description
For use with vacuum filtration systems or bottle top vacuum filters
Specifications
Specifications
150 mL filter funnels, Corning bottle top and vacuum filtration systems
Non-sterile
100/Pk.
Fiberglass
100
Prefilter