Learn More
Corning™ Mediatech™ Cell Culture Phosphate Buffered Saline (10X)
With low endotoxin content
Brand:  Corning™ 46-013-CM
Additional Details : Weight : 3.60000kg
Product Code. 15316257
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Maintains cell culture media in the physiological pH range
  • Maintains cell tonicity and viability
  • Produced under the highest industry standards to ensure superior results
  • Nontoxic
  • Liquid without calcium and magnesium
Specifications
Specifications
No Calcium, No Magnesium
10 X
Potassium Chloride 2g/L, Potassium Dihydrogen Phosphate 2.40g/L, Sodium Chloride 80g/L, Disodium Phosphate 14.4g/L
Liquid
15° to 30°C
6 x 1000 mL
7.3 to 7.5