Learn More
Corning™ Mediatech™ Cell Culture Phosphate Buffered Saline (10X)
With low endotoxin content
Brand: Corning™ 46-013-CM
Additional Details : Weight : 3.60000kg
Description
- Maintains cell culture media in the physiological pH range
- Maintains cell tonicity and viability
- Produced under the highest industry standards to ensure superior results
- Nontoxic
- Liquid without calcium and magnesium
Specifications
No Calcium, No Magnesium | |
10 X | |
Potassium Chloride 2g/L, Potassium Dihydrogen Phosphate 2.40g/L, Sodium Chloride 80g/L, Disodium Phosphate 14.4g/L | |
Liquid |
15° to 30°C | |
6 x 1000 mL | |
7.3 to 7.5 |