Learn More
Corning™ CellCube™ Tubing Adaptor
A new closed system offering designed to help reduce the risk of adventitious contamination
340.70 EUR - 1249.25 EUR
Specifications
Diameter (Metric) Inner Tubing | 9.53 mm |
---|---|
No. per Case | 4/Cs. |
No. per Pack | 1/Pk. |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
17249109
|
Corning™
3236 |
4 AseptiQuik™ S Connectors, 4 Tubes Clamps |
1249.25 EUR
Pack of 4 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
17259109
|
Corning™
3237 |
AseptiQuik™ S Connected to a Female MPC |
340.70 EUR
Pack of 4 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
17269109
|
Corning™
3238 |
AseptiQuik™ S Connected to a Male MPC |
340.70 EUR
Pack of 4 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
Description
- Closed System Cell Cube
- CellCube 10, 25, 100 modules, circulation loops, and connectors
- Allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik™ and MPC connectors.
Specifications
9.53 mm | |
4/Cs. | |
1/Pk. |