missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) nitrate trihydrate, 99%, pure, Thermo Scientific Chemicals

Catalog Number p-7077452
Click to view available options
Quantity:
1 kg
5 kg
10031-43-3
CuN2O6·3H2O
241.6
SXTLQDJHRPXDSB-UHFFFAOYSA-N
copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem
9837674
copper;dinitrate;trihydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]
10031-43-3
CuN2O6·3H2O
241.6
SXTLQDJHRPXDSB-UHFFFAOYSA-N
copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem
9837674
copper;dinitrate;trihydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 114.0°C
Quantity 1 kg
Assay Percent Range 98.5 to 104% (Iodometry)
Linear Formula Cu(NO3)2·3H2O
Merck Index 15,2633
Solubility Information Solubility in water: 267 g/100 mL. Other solubilities: freely soluble in alcohol, practically insoluble in ethyl acetate
Formula Weight 241.6
Percent Purity 99%
Grade Pure
Chemical Name or Material Copper(II) nitrate trihydrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) nitrate, trihydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'