missing translation for 'onlineSavingsMsg'
Learn More

Copper(Ii) Nitrate Hemi(Pentahydrate), Honeywell Fluka™

Catalog Number p-7071789
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 1 kg
This item is not returnable. View return policy
19004-19-4
MFCD02253001
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate
This item is not returnable. View return policy
19004-19-4
MFCD02253001
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate

≥98%,≥26% Cu basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 500 g
Linear Formula Cu(NO3)2 · 2.5H2O
UN Number 3085
Formula Weight 232.59g/mol
Percent Purity ≥98%
Packaging Plastic Bottle
Chemical Name or Material Copper(Ii) Nitrate Hemi(Pentahydrate)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'