Learn More
SI Analytics™ Conductivity Cells
SI Analytics™ Conductivity Cells are trace conductivity cells with an integrated flow-through vessel.
Brand: SI Analytics™ 285414360
Additional Details : Weight : 0.50000kg
Description
- Trace conductivity cell
- Includes cable and 8-pole plug
- Available with ID function
Specifications
0°C to 100°C | |
Conductivity Cell | |
120 mm | |
Stainless Steel Sensor | |
Fixed cable, 8-pole plug, and ID function |
0.01 cm-1 | |
12 mm | |
1.5 m | |
Up to 0.3 mS/cm | |
LF 213 T ID |