Learn More
Ohaus™ CSE Series Compact Scales
Designed for educational weighing applications. Ohaus™ CSE Series Compact Scales are the ideal choice for economical and accurate educational applications with capacities up to 5kg.
45.00 EUR
Specifications
Material ABS housing
Electrical Requirements AC adapter
Display Type LCD
Certifications/Compliance OIML M1 or ASTM Class F
View More Specs
Products 2
Product Code Brand Model Capacity Readability Stabilization Time Price Quantity & Availability  
Product Code Brand Model Capacity Readability Stabilization Time Price Quantity & Availability  
12747236 View Documents Ohaus™
80500630
CS200E 200 g 0.1 g 3 sec. 45.00 EUR
Each View alternative products
Save Up to 
»
Estimated Shipment
17-07-2019
Add to basket
12757236 View Documents Ohaus™
80500631
CS2000E 2 kg (72 oz.) 1 g 3 sec. 45.00 EUR
Each View alternative products
Save Up to 
»
Estimated Shipment
17-07-2019
Add to basket
Description
Description
  • Liquid crystal display (LCD)
  • AC adapter (not included) or 3 AA batteries (included)
  • ABS Housing, Stainless Steel Pan
  • Mechanical and software overload/underload protection, auto shut-off
Specifications
Specifications
ABS housing
AC adapter
LCD
OIML M1 or ASTM Class F