Learn More
Buerkle™ HDPE, Compact Jerry Can (Electroconductive) with Threaded Connector
Catalog Number 16319541
Click to view available options
Capacity (Metric):
5 L
10 L
For Use With: For use in explosion-proof areas; Closure Material: HDPE; Color: Black
  • HDPE, black, anti-static
  • For use in explosion-proof areas
  • For dispensing and storing combustible and highly flammable liquids
  • With or without threaded connector 3/4 in.
  • Due to its narrow width, the canister requires very little room
Capacity (English) 1.3 gal.
Product Type Jerrican
Closure Color Black
Closure Material HDPE
Description Canister
Capacity (Metric) 5 L
Dimensions (L x W) 335 x 65 mm
Dimensions (L x W x H) 335 x 65 x 335 mm
Material Anti-static HDPE
Disposable No
For Use With (Application) For Use in Explosion-Proof Areas
Graduations Liter
Height (English) 13.19 in.
Height (Metric) 335 mm
Length (English) 13.19 in.
Length (Metric) 335 mm
No. per Pack 1
Width (English) 2.56 in.
Width (Metric) 65 mm
Color Black
For Use With (Equipment) StopCock PA