Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 1mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11857603
  • 19.70 EUR / Each
Estimated Shipment: 15-07-2024
Description
  • Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
  • Supplied as standard
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Mini SDS PAGE, Native PAGE, Gradient, Second Dimension and Nucleic Acid Separations
Comb
1 Each
20 μL
OmniPAGE Mini Vertical Gel Electrophoresis Unit
20
1 mm