missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Comb for Horizontal Tank

For use with Horizontal Tank

Brand:  Fisherbrand™

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11740996

Quantity Price
1 71.83 EUR / Each
10 62.03 EUR / Each
Estimated Shipment
22-03-2021
Add to basket


Description

Description

  • Color code according to thickness
  • Volume per well given for deposit height of 1mm
  • Well spacing of μT combs is compatible with multichannel pipettes
Specifications

Specifications

Clear
Horizontal Tank Varige
2.4 μL
Thickness: 1mm
Comb
15 cm
Videos
SDS
Documents