Learn More
Fisherbrand™ Comb 1.5mm
Manufactured to the highest standard. Fisherbrand™ Comb 1.5mm is used with electrophoresis systems.
Brand: Fisherbrand™
Additional Details : Weight : 0.80000kg
Description
- Used with vertical electrophoresis systems
Specifications
Vertical Gel Electrophoresis | |
10 | |
1.5 mm |