Learn More
Shamrock™ Color Coded Multi-Purpose 3 Inch Labeling Tape
Offered in a number of colors to fit your needs. Shamrock™ Color Coded Multi-Purpose 3 Inch Labeling Tape offers multiple opportunities for color coding in the laboratory - test, physician or department.
Brand:  Shamrock™ ST-340-14
Additional Details : Weight : 0.13770kg
 View more versions of this product
Product Code. 11790085
  • 27.50 EUR / 55m
Estimated Shipment: 21-05-2024
Description
  • Resistant to water, oil and acid
  • Temperature range of -20 to +121°C
  • For autoclave, freezer, refrigerator and oven
  • Universal marking with ballpoint pen, paper pencil and felt marker
Specifications
Specifications
Removable
Color Coded Multi-Purpose Labeling Tape
55 m
Gray
55 m x 19 mm
19 mm