missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cobalt(II) molybdenum oxide hydrate, 99.9% (metals basis)

Brand:  Alfa Aesar™ 045860.09

Additional Details : CAS Number : 13762-11-3 Weight : 0.01000kg Transport : UN number : 3077 Chem class : 9 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 15447115

  • 42.40 EUR / 10g


Estimated Shipment date
from Supplier 24-04-2019
Add to basket

Specifications

Chemical Identifiers

13762-11-3
236.894
QPJIKZNJXISODY-UHFFFAOYSA-N
44135796
O.[O-][Mo](=O)(=O)[O-].[Co+2]
CoH2MoO5
MFCD00150209
cobalt 2+ dioxido dioxo molybdenum-water 1/1/1, cobaltic molybdate, co.moo4.h2o, cobalt molybdate monohydrate, cobalt 2+ hydrate molybdate, cobalt ii molybdenum oxide hydrate
cobalt(2+);dioxido(dioxo)molybdenum;hydrate

Specifications

Cobalt (II) molybdenum oxide hydrate
(metals basis)
MFCD00150209
cobalt 2+ dioxido dioxo molybdenum-water 1/1/1, cobaltic molybdate, co.moo4.h2o, cobalt molybdate monohydrate, cobalt 2+ hydrate molybdate, cobalt ii molybdenum oxide hydrate
O.[O-][Mo](=O)(=O)[O-].[Co+2]
236.894
218.87 (Anhydrous)
99.9%
Purple
13762-11-3
CoH2MoO5
UN3077
QPJIKZNJXISODY-UHFFFAOYSA-N
cobalt(2+);dioxido(dioxo)molybdenum;hydrate
44135796
Powder
−325 Mesh
10g
Videos
SDS