Learn More
Thermo Scientific™ ClipTip™ Pipette Tip Racks
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
Brand: Thermo Scientific™ 94410619
27.60 EUR valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Additional Details : Weight : 0.95000kg
Description
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
- Small Rack: For use with ClipTip 20 and 50 Pipette Tips
- Medium Rack: For use with ClipTip 200 and 300 Pipette Tips
- Large Rack: For use with ClipTip 1000 and 1250 Pipette Tips
- CT300 Ext Rack: For use with ClipTip 300 Ext Pipette Tips
Ordering Information:
Racks are supplied without tips and inserts
Specifications
ClipTip Empty Rack | |
ClipTip Empty Rack | |
ClipTip 300 Ext Pipette Tip | |
ClipTip |