Learn More
Nickel Electro™ Clifton™ Stainless Steel Tank
Stainless Steel Tank providing an external vessel of circulating temperature controlled water.
Brand:  Nickel Electro™ NE4-8U
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11756587
  • 280.00 EUR / Each
Estimated Shipment
04-03-2021
Add to basket
Description
Description
  • Double-walled construction
  • External surfaces have an anti-bacterial paint finish
  • Inner tank is a bright stainless steel.
  • While in use, submerge Inlet and outlet hoses under water from NE6-FH Clifton Flow Heater.
Specifications
Specifications
Laboratory use
Stainless Steel, Antibacterial Paint Finish
8 L
Clifton Thermostirrer
Stainless Steel Tank