Learn More
Nickel Electro™ Clifton™ Centrifuge Rotor Angle Head
Nickel Electro™ Clifton™ Centrifuge Rotor Angle Head comes in different capacities that hold either 8, 12, or 24 tubes.
POA
Specifications
For Use With (Equipment) Universal Rotor
View More Specs
Includes: dimCapacity
Description
Description
  • Can be removed from centrifuge for cleaning
  • The advantage of the angle head is that tubes which have large screw caps can be centrifuged.
  • The large capped tubes obstruct the normal swing out head.
  • Converts normal swing out head range Centrifuge to a fixed head.
Universal rotor, angle head, fixed head
Specifications
Specifications
Universal Rotor