Learn More
Thermo Scientific™ Cleaning Kit for Laboratory Crucibles
For safe cleaning of crucibles without damage
Brand: Thermo Scientific™ 037995.KT
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Reduces sample contamination
- Includes liquid cleaner, variety of abrasive materials and instructions
- For cleaning most lab crucibles, including metallic crucibles such as zirconium, nickel, molybdenum, tantalum and platinum, and crucibles of ceramic or glass