missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-2-(Boc-amino)cyclopropanemethanol, 97%, Thermo Scientific Chemicals
301.34 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
cis-2-(Boc-amino)cyclopropanemethanol is employed as a intermediate for chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-2-(Boc-amino)cyclopropanemethanol is employed as a intermediate for chemical research.
Notes
Keep container tightly closed. Store away from oxidizing agent.
cis-2-(Boc-amino)cyclopropanemethanol is employed as a intermediate for chemical research.
Notes
Keep container tightly closed. Store away from oxidizing agent.
Specifications
Formula Weight | 187.23 |
Percent Purity | 97% |
Quantity | 250 mg |
Chemical Name or Material | cis-2-(Boc-amino)cyclopropanemethanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'