missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ cis-2-(Boc-amino)cyclohexanecarboxylic acid, 97%

Brand:  Alfa Aesar™ H62600.06

Additional Details : CAS Number : 63216-49-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15459108

  • 429.00 EUR / 5g


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

63216-49-9
243.303
QJEQJDJFJWWURK-BDAKNGLRSA-N
1512487
CC(C)(C)OC(=O)NC1CCCCC1C(=O)O
C12H21NO4
MFCD01863244
1r,2s-boc-achc, boc-1,2-cis-achc-oh, boc-cis-2-aminocyclohexanecarboxylic acid, cis-2-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,2s-boc-2-aminocyclohexane carboxylic acid, n-boc-+/--cis-2-amino-cyclohexane-carboxylic acid, cis-2-tert-butoxycarbonylamino-cyclohexanecarboxylic acid, 1r,2s-2-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, cis-1-t-butyloxycarbonylamino-cyclohexyl-2-carboxylic acid, 1r,2s-2-tert-butoxy carbonyl amino cyclohexane-1-carboxylic acid
(1R,2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid

Specifications

cis-2-(Boc-amino)cyclohexanecarboxylic acid
5g
C12H21NO4
1r,2s-boc-achc, boc-1,2-cis-achc-oh, boc-cis-2-aminocyclohexanecarboxylic acid, cis-2-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,2s-boc-2-aminocyclohexane carboxylic acid, n-boc-+/--cis-2-amino-cyclohexane-carboxylic acid, cis-2-tert-butoxycarbonylamino-cyclohexanecarboxylic acid, 1r,2s-2-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, cis-1-t-butyloxycarbonylamino-cyclohexyl-2-carboxylic acid, 1r,2s-2-tert-butoxy carbonyl amino cyclohexane-1-carboxylic acid
CC(C)(C)OC(=O)NC1CCCCC1C(=O)O
243.303
243.29
97%
127°C to 131°C
63216-49-9
MFCD01863244
QJEQJDJFJWWURK-BDAKNGLRSA-N
(1R,2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
1512487
Solid
Videos
SDS