Learn More
Thermo Scientific Chemicals Ciprofloxacin hydrochloride hydrate, 98%
Catalog Number p-7077646
Click to view available options
Quantity:
5 g
25 g
128074-72-6
C17H19ClFN3O3
367.81
MFCD00242856
DIOIOSKKIYDRIQ-UHFFFAOYSA-N
62998
CHEBI:59936
1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride
Cl.OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
128074-72-6
C17H19ClFN3O3
367.81
MFCD00242856
DIOIOSKKIYDRIQ-UHFFFAOYSA-N
62998
CHEBI:59936
1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride
Cl.OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
Ciprofloxacin hydrochloride hydrate, CAS # 128074-72-6, is the hydrate form of the hydrochloride salt of ciprofloxacin, a broad-spectrum antibiotic.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Ciprofloxacin HCl is a fluoroquinolone antibiotic that works by binding to bacterial DNA gyrase/DNA topoisomerase, thus inhibiting DNA replication and subsequent bacterial cell division
• It can cause double-stranded breaks in the bacterial chromosome
• Ciprofloxacin HCl is an analog of the antibiotic nalidixic acid
• Works as an antibiotic against both gram-positive and gram-negative bacteria and can have fungicidal activity
Applications
• Used for elimination of mycoplasma in cell culture
• Can be used to study antimicrobial activity and resistance in laboratory settings
TRUSTED_SUSTAINABILITY
Type Ciprofloxacin Hydrochloride Hydrate
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Formula Weight 367.81
Percent Purity 98%
Water 4.0 to 7.0%
Quantity 5 g
Chemical Name or Material Ciprofloxacin hydrochloride hydrate