Learn More
Proteintech ChromoTek Dnmt1-Chromobody™ Plasmid (TagRFP)
Dnmt1-Chromobody™ fused to TagRFP.
Brand:  Proteintech DCR
Additional Details : Weight : 0.00240kg
Product Code. 17373283
  • 874.00 EUR / 20µg
Estimated Shipment: 04-06-2024
Description
  • DNA plasmid encoding for anti-Dnmt1 VHH (anti-Dnmt1 Nanobody) fused to TagRFP
Specifications
Specifications
20 μg