Learn More
Proteintech ChromoTek Actin-Chromobody™ Plasmid (TagGFP2)
Actin-Chromobody™ fused to TagGFP2.
Brand:  Proteintech ACG
Additional Details : Weight : 0.00240kg
Product Code. 17303283
  • 874.00 EUR / 20µg
Estimated Shipment: 04-06-2024
Description
  • DNA plasmid encoding for anti-Actin VHH (anti-Actin Nanobody) fused to TagGFP2
Specifications
Specifications
20 μg