Learn More
Macherey-Nagel™ Chromofil™ Polyester Syringe Filter
Filter polar or nonpolar solvents. Chromofil™ PET Polyester Syringe Filter is hydrophilic for multipurpose applications and comes color coded.
Brand:  Macherey-Nagel™ 729020
Additional Details : Weight : 0.33300kg
 View more versions of this product
Product Code. 10399860
  • 142.42 EUR / Pack of 100
Estimated Shipment: 02-08-2024
Description
  • Syringe filters with polyester membrane  
  • Color-coded 
  • Hydrophilic multipurpose membrane
  • For polar as well as nonpolar solvents
  • The HPLC filter, especially suited for mixtures of water and organic solvents 
  • For TOC/DOC determination
  • Not cytotoxic, does not inhibit the growth of microorganisms and higher cells
Specifications
Specifications
Colorless Top and Orange Bottom
0.45 μm
Hydrophilic
Syringe Filter
Polyester
Non Polar and Polar Solvent
100 Pack
25 mm