Learn More
Macherey-Nagel™ Chromafix™ SB Cartridge
191.00 EUR - 266.00 EUR
Specifications
Base Material | Silica |
---|---|
For Use With (Application) | Organic Acids, Caffeine, Saccharin |
Particle Size | 45 μm |
Sorbent | SB |
Type | Chromafix SB Cartridge |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10236571
|
Macherey-Nagel™
731836 |
Adsorbent Weight: 920 mg |
266.00 EUR
Pack of 50 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
10053051
|
Macherey-Nagel™
731835 |
Adsorbent Weight: 460 mg |
200.00 EUR
Pack of 50 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
10458981
|
Macherey-Nagel™
731834 |
Adsorbent Weight: 230 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Silica | |
45 μm | |
Chromafix SB Cartridge | |
50 Pack |
Organic Acids, Caffeine, Saccharin | |
SB | |
60 Å | |
500 m2/g |