Learn More
Chlorthalidone, MedChemExpress
Product Code. p-200098913
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16446858
Brand: Medchem Express HY15833
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
MedChemExpress Chlorthalidone is a thiazide-like diuretic used to treat hypertension.
Chlorthalidone is a thiazide-like diuretic used to treat hypertension.
Specifications
Chemical Name or Material Chlorthalidone
CAS 77-36-1
Color White
Purity Grade Notes Research
Molecular Formula C14H11ClN2O4S
Quantity 10 mg
Solubility Information DMSO : ≥ 41 mg/mL (121.03 mM)
SMILES O=S(C1=CC(C2(O)NC(C3=C2C=CC=C3)=O)=CC=C1Cl)(N)=O
Molecular Weight (g/mol) 338.77
Formula Weight 338.77
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation