missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorotris(triphenylphosphine)cobalt(I), 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
A stoichiometric reducing agent that is used in the radical dimerization of halogenated organic molecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A stoichiometric reducing agent that is used in the radical dimerization of halogenated organic molecules.
Solubility
Insoluble in water.
Notes
Air & Moisture Sensitive.
A stoichiometric reducing agent that is used in the radical dimerization of halogenated organic molecules.
Solubility
Insoluble in water.
Notes
Air & Moisture Sensitive.
Specifications
Melting Point | 135°C to 139°C |
Quantity | 1 g |
Sensitivity | Air and moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 881.26 |
Percent Purity | 97% |
Chemical Name or Material | Chlorotris(triphenylphosphine)cobalt(I) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'