missing translation for 'onlineSavingsMsg'
Learn More

Chloromethylphosphonic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-4813403
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2565-58-4
CH4ClO3P
130.46
MFCD00013955
MOFCYHDQWIZKMY-UHFFFAOYSA-N
chloromethyl phosphonic acid, phosphonic acid, chloromethyl, chloromethanephosphonic acid, phosphonic acid, p-chloromethyl, monochloromethyl phosphonic acid, chloromethylphosphite, phosphonomethyl chloride, chloromethyl-phosphonicaci, phosphonic acid,p-chloromethyl, chloromethyl phosphonic acid, 98 %
75723
chloromethylphosphonic acid
OP(O)(=O)CCl
This item is not returnable. View return policy
2565-58-4
CH4ClO3P
130.46
MFCD00013955
MOFCYHDQWIZKMY-UHFFFAOYSA-N
chloromethyl phosphonic acid, phosphonic acid, chloromethyl, chloromethanephosphonic acid, phosphonic acid, p-chloromethyl, monochloromethyl phosphonic acid, chloromethylphosphite, phosphonomethyl chloride, chloromethyl-phosphonicaci, phosphonic acid,p-chloromethyl, chloromethyl phosphonic acid, 98 %
75723
chloromethylphosphonic acid
OP(O)(=O)CCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 88.0°C to 92.0°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 130.47
Percent Purity 98%
Chemical Name or Material Chloromethylphosphonic acid
compliance-icons
Product Identifier
  • Chloromethylphosphonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'