missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Chloromethyl Latex Beads, 4% w/v, 0.4 μm
Chloromethyl Latex Beads, 4% w/v, 0.4 μm
Brand: Molecular Probes™ C37346
Additional Details : Weight : 0.00525kg
Description
Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Because of the way that the polystyrene chains arrange themselves in the bead, the surface is very hydrophobic in character, making these ideal materials for the adsorption of materials such as proteins.The chloromethyl latex has a high density of chloromethyl groups attached to the styrene monomeric unit. These reactive surface functional groups react directly with amino groups in antibodies, antigens, or other ligands under mild aqueous conditions to yield a stable covalent product by a one-step process. The particles are stabilized by negatively-charged sulfate groups. This type of particle can be used at both high and low pH conditions. This is a hydrophobic latex.
Microspheres Are Available in Many Sizes and Surface Functionalities
- 25 sizes to choose from—0.02 microns to 16 microns
- 20 different surface—for ready conjugation to virtually any material
- Surfactant free—ultraclean
- Various quantities—from milliliter to 500-liter amounts
Key Applications of Microspheres - Instrument calibration (flow cytometry, microscopy, HTS, HCS)
- Flow testing (microfluidics, blood flow, water flow, and air flow)
- Cell biology tracers (cell differentiation and cell tracing)
- Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)
For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
Specifications
Store in refrigerator (2–8°C). | |
Latex Beads | |
Room Temperature | |
0.4 μm |
Polystyrene | |
IDC | |
Chloromethyl |
For Research Use Only. Not for use in diagnostic procedures.