Learn More
WTW™ Chlorine Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Marke: WTW™ 251414
Weitere Details : Weight : 0.80000kg
Spezifikation
DIN 38408 G4, EN ISO 7393 | |
Disinfectants, wastewater, drinking water | |
0.02 to 2.00 mg/L Cl2, total | |
4-Dimethylaminobenzaldehyde |
5 mL | |
PhotoLab™ 6000 photometer, Photoflex™ meter | |
15°C to 25°C | |
Powder Pillows Kit |