missing translation for 'onlineSavingsMsg'
Learn More

Chloride Standard Solution, WTW™

Catalog Number p-8014355
Click to view available options
Quantity:
500 mL
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Standard solutions to calibrate contains 10 g of the ion to be measured per 10 liters. By defined dilutions of these solutions, the user can easily produce his own calibration solution for the desired measuring range. The bulk with a one liter content makes for economic measurements.

For few parameters such as nitrite, which are subject to a quick conversion, freshly prepared control standards of stock solutions versus the multi-parameter standard checks are mandatory. Stock solutions are also more cost-effective than the already confectioned CombiChecks™. With stock solutions, the correct dilution must be self-calculated and conducted. The use of standards is supported via AQA functions in the photoLab™ photometers.
  • Free chlorine
  • Bound chlorine
  • Formaldehyde
  • Hydrazine
  • Hydrogen peroxide
  • Hydrogen sulfide
  • Phenol
  • Silicon
  • Sulfide
  • Sulfite
  • Anionic surfactants
Physical Form Liquid
Concentration 1000mg/L
Solution Contains Chloride
Traceability to NIST Yes
pH 0.5
Boiling Point 100°C
Color Colorless
Quantity 500 mL
Solubility Soluble in water
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'