missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine Free Base, >85%, MP Biomedicals™

Catalog Number p-4679702
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
55-56-1
C22H30Cl2N10
505.452
MFCD00009673
GHXZTYHSJHQHIJ-UHFFFAOYSA-N
chlorhexidine, chlorhexidinum, chlorhexidin, nolvasan, rotersept, tubulicid, fimeil, hexadol, soretol, cloresidina
9552079
CHEBI:3614
(1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine
C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl
This item is not returnable. View return policy
55-56-1
C22H30Cl2N10
505.452
MFCD00009673
GHXZTYHSJHQHIJ-UHFFFAOYSA-N
chlorhexidine, chlorhexidinum, chlorhexidin, nolvasan, rotersept, tubulicid, fimeil, hexadol, soretol, cloresidina
9552079
CHEBI:3614
(1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine
C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl
Molecular biology; Cell biology
Color White
Quantity 5 g
Physical Form Solid
Assay Percent Range >85%
Formula Weight 505.5
Chemical Name or Material Chlorhexidine Free Base, Free Base
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'