Learn More
Hanna Instruments™ Chemical Test Kit
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.
Brand: Hanna Instruments™ HI38001
Additional Details : Weight : 0.50000kg
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
- Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
- High resolution readings for fast, accurate determinations
- Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
Barium chloride | |
Water samples | |
100 ea. (200 total) | |
100 to 1000mg/L and 1,000 to 10,000mg/L | |
Titration |
Sulfate determination | |
100 packets sulfate reagent A (2 sets), 100mL LR sulfate reagent B, 100mL HR sulfate reagent B, 10mL sulfate reagent C, 20mL complexing agent, 30mL sulfate solution, 50mL plastic vessels (2), and 1mL syringes (2) | |
1 Kit | |
High range and low range determinations | |
Chemical Test Kit |