Learn More
Hanna Instruments™ Chemical Test Kit
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.
Brand: Hanna Instruments™ HI3875
Additional Details : Weight : 0.50000kg
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
- Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
- High resolution readings for fast, accurate determinations
- Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
DPD | |
Water samples | |
100 | |
0.0 to 3.5mg/L | |
Titration |
Free chlorine MR determination | |
HI93701-0 free chlorine reagent (100 packets), 500mL deionized water, checker disc, glass vials with caps (2), and 3mL plastic pipette | |
1 Kit | |
Medium range determination | |
Chemical Test Kit |