Learn More
Hanna Instruments™ Chemical Test Kit
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.
Brand: Hanna Instruments™ HI3833
Additional Details : CAS Number : 7790-62-7 Weight : 0.16000kg Transport : UN number : 2923 Chem class : 8 Pack group : II
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
- Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
- High resolution readings for fast, accurate determinations
- Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
Ascorbic acid | |
Water Samples | |
50 | |
0 to 5mg/L | |
Phosphate Test kit |
Phosphate determination | |
20mL plastic beaker, color comparison cube, and 50 packets phosphate reagent | |
1 Kit | |
Titration |