missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride, 99.99%, (Optical Grade I), MP Biomedicals

Catalog Number p-3547058
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
This item is not returnable. View return policy
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
24293
CHEBI:63039
caesium(1+) chloride
[Cl-].[Cs+]
This item is not returnable. View return policy
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
24293
CHEBI:63039
caesium(1+) chloride
[Cl-].[Cs+]

Solutions are clear and colorless suitable for density gradient work, nucleic acid and protein purification

Quantity 25 g
Assay Percent Range 99.99%
Formula Weight 168.4
Grade Optical I
Chemical Name or Material Cesium Chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'