missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium(IV) sulfate, 0.1N Standardized Solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
4 L
Description
Cerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions' reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 1 L |
Color | Yellow |
Formula Weight | 332.24 |
Odor | Odorless |
Density | 1.05 |
Solubility | Miscible with water and dilute sulfuric acid |
Physical Form | Liquid |
Chemical Name or Material | Cerium(IV) sulfate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'