Learn More
Macherey-Nagel™ Cellulose Mn
107.00 EUR - 475.00 EUR
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10194611
|
Macherey-Nagel™
816250.5 |
5 kg |
475.00 EUR
5kg |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
10134571
|
Macherey-Nagel™
816250.1 |
1 kg |
107.00 EUR
1kg |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
Specifications
Cellulose Mn | |
Cellulose |