Learn More
BINDER™ CELLROLL Roller Bottle System
For Use With: BINDER units CB 220
Brand: BINDER™ 8012-0572
Additional Details : Weight : 5.00000kg
Description
- Modular
- Expandable
- For either 4 or 6 roller bottles
Specifications
CELLROLL Set - modular, expandable roller bottle system for cell cultivation, 6 roller bottles | |
6 roller bottles | |
Cell cultivation |
BINDER Units CB 220 | |
Roller Bottle System |