Learn More
Cefotetan, 95+%, Thermo Scientific Chemicals
Catalog Number p-7077802
Click to view available options
Quantity:
250 mg
1 g
69712-56-7
C17H17N7O8S4
575.63
SRZNHPXWXCNNDU-RHBCBLIFSA-N
cefotetan, cefotetanum, apacef, cefotetanum inn-latin, 6r,7s-7-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, cefotan tn, 6r,7s-7-4-carbamoylcarboxymethylene-1,3-dithiethane-2-carboxamido-7-methoxy-3-1-methyl-1h-tetrazol-5-yl thio methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, 7beta-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7alpha-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-3,4-didehydrocepham-4-carboxylic acid, 6r,7s-7-4-1-amino-3-hydroxy-1,3-dioxopropan-2-ylidene-1,3-dithietane-2-carbonyl amino-7-methoxy-3-1-methyltetrazol-5-yl sulfanylmethyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid
53025
(6R,7S)-7-[[4-(2-amino-1-carboxy-2-oxoethylidene)-1,3-dithietane-2-carbonyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C4SC(=C(C(=O)N)C(=O)O)S4)OC)SC2)C(=O)O
69712-56-7
C17H17N7O8S4
575.63
SRZNHPXWXCNNDU-RHBCBLIFSA-N
cefotetan, cefotetanum, apacef, cefotetanum inn-latin, 6r,7s-7-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, cefotan tn, 6r,7s-7-4-carbamoylcarboxymethylene-1,3-dithiethane-2-carboxamido-7-methoxy-3-1-methyl-1h-tetrazol-5-yl thio methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, 7beta-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7alpha-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-3,4-didehydrocepham-4-carboxylic acid, 6r,7s-7-4-1-amino-3-hydroxy-1,3-dioxopropan-2-ylidene-1,3-dithietane-2-carbonyl amino-7-methoxy-3-1-methyltetrazol-5-yl sulfanylmethyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid
53025
(6R,7S)-7-[[4-(2-amino-1-carboxy-2-oxoethylidene)-1,3-dithietane-2-carbonyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C4SC(=C(C(=O)N)C(=O)O)S4)OC)SC2)C(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: 0.51 mg/mL. Other solubilities: slightly soluble in methanol
Formula Weight 575.63
Percent Purity 95+%
Chemical Name or Material Cefotetan
compliance-icons
Product Identifier
  • Cefotetan
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .