missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK5/p25 Recombinant Human Protein
CDK5 plays a critical role in neuron development. This protein is coexpressed and copurified with p25.
1826.00 EUR - 7890.00 EUR
Specifications
Accession Number | NP_004926.1 I NP_003876 |
---|---|
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 87.7kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468673
|
Invitrogen™
PR8543B |
100μg |
1826.00 EUR
100µg |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15488743
|
Invitrogen™
PV4677 |
1mg |
7890.00 EUR
1mg |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The disregulation of the CDK5 and the truncated activator p25 is believed to be involved in neurodegenerative diseases such as Alzheimer's, Parkinson's, and Amyotrophic Lateral Sclerosis.
Specifications
NP_004926.1 I NP_003876 | |
87.7kDa | |
RUO - research use only | |
CDK5/p25, CDK5R | |
Human | |
GST Tag, His Tag | |
Frozen |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CDK and CHK Kinase Family | |
Recombinant | |
Baculovirus |