missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99/MIC2 (Ewing' s Sarcoma Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™
Detect CD99 in human and rat samples with Epredia™ CD99/MIC2 (Ewing's Sarcoma Marker) Ab-1, Mouse Monoclonal Antibody, which provides accurate, reproducible results.
193.00 EUR - 565.00 EUR
Specifications
Antigen | CD99/MIC2 (Ewings Sarcoma Marker) Ab-1 |
---|---|
Clone | HO36-1.1 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
12680837
|
Epredia
MS-294-P0 |
Concentrated | 200μg/mL | 100 μL |
193.00 EUR
100µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12690837
|
Epredia
MS-294-P1 |
Concentrated | 200μg/mL | 500 μL |
545.00 EUR
500µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12670837
|
Epredia
MS-294-P |
Concentrated | 200μg/mL | 1 mL |
565.00 EUR
1mL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12640847
|
Epredia
MS-294-R7 |
Prediluted | — | 7 mL |
193.00 EUR
7mL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12610847
|
Epredia
MS-294-PABX |
Concentrated | 1mg/mL | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12600847
|
Epredia
MS-294-P1ABX |
Concentrated | 1mg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
CD99, or MIC2 gene product, or E2 antigen is expressed on the cell membrane of some lymphocytes, cortical thymocytes, and granulosa cells of the ovary. The antigen is also expressed by most pancreatic islet cells, Sertoli cells of the testis, and some endothelial cells. Mature granulocytes express very little or no CD99. MIC2 is strongly expressed on Ewing's sarcoma cells and primitive peripheral neuroectodermal tumors.Host Species: Mouse
Clone: HO36-1.1
Isotype: IgM
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Purified E-rosette forming cells from human peripheral blood lymphocytes
Molecular Weight: 27-32kDa
Positive Control: Pancreas or Ewing's sarcoma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
CD99/MIC2 (Ewings Sarcoma Marker) Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human, Rat | |
IgM |
HO36-1.1 | |
Monoclonal | |
Mouse | |
Oncogenesis | |
Purified E-rosette forming cells from human peripheral blood lymphocytes | |
Primary |