missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90.2 Monoclonal Antibody (30-H12), PE, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen A14729
Additional Details : Weight : 0.01000kg
Description
CD90.2 Monoclonal antibody specifically detects CD90.2 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD90.2 | |
Monoclonal | |
0.1 mg/mL | |
PBS with <0.1% sodium azide | |
P01831 | |
Thy1 | |
Mouse thymus or spleen | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b κ |
Flow Cytometry | |
30-H12 | |
PE | |
Thy1 | |
CD90; FLJ33325; T25; Thy 1.2; Thy1; Thy-1; Thy-1 antigen; thy-1 membrane glycoprotein; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta | |
Rat | |
purified | |
RUO | |
21838 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |